Learn More
2,5-Difluoro-4-hydroxybenzaldehyde, 99%
Product Code. 11345145
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
Product Code. 11345145
Brand: Thermo Scientific Alfa Aesar H31528.03
Request Bulk or Custom Format
CAS: 918523-99-6 | C7H4F2O2 | 158.104 g/mol
2,5-Difluoro-4-hydroxybenzaldehyde is used as primary and secondary intermediate and as pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2,5-Difluoro-4-hydroxybenzaldehyde is used as primary and secondary intermediate and as pharmaceutical intermediate.
Solubility
Slightly soluble in water.
Notes
Air sensitive. Store away from oxidizing agents, air. Store under inert gas.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 918523-99-6
Molecular Formula C7H4F2O2
Molecular Weight (g/mol) 158.104
MDL Number MFCD04112521
InChI Key FFPHRYLGIPAZBL-UHFFFAOYSA-N
Synonym 4-hydroxy-2,5-difluorobenzaldehyde, 2,5-difluoro-4-hydroxy-benzaldehyde, benzaldehyde, 2,5-difluoro-4-hydroxy, ksc496a0b
PubChem CID 21966462
IUPAC Name 2,5-difluoro-4-hydroxybenzaldehyde
SMILES C1=C(C(=CC(=C1F)O)F)C=O
Specifications
Melting Point 145°C to 148°C
Quantity 1 g
Sensitivity Air Sensitive
Solubility Information Slightly soluble in water.
Formula Weight 158.11
Percent Purity 99%
Chemical Name or Material 2,5-Difluoro-4-hydroxybenzaldehyde