Learn More
2,5-Difluorobenzyl bromide, 98%
Product Code. 11485616
Change view
Click to view available options
Quantity:
2.5 g
Unit Size:
2.5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11485616 2.5 g 2.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11485616 Supplier Thermo Scientific Alfa Aesar Supplier No. B20752.88
 Restricted Item
Estimated Shipment: 29-12-2026
Add to basket
missing translation for 'validMainframeMsg'
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Specifications
Quantity 2.5 g
Percent Purity 98%
Physical Form Liquid