missing translation for 'onlineSavingsMsg'
Learn More

2,5-Dihydroxy-1,4-benzoquinone, TRC

Product Code. 30388480
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
Product Code. Quantity unitSize
30388480 1 g 1g
1 options
This item is not returnable. View return policy

Product Code. 30388480

Brand: Toronto Research Chemicals TRCD4471051G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2,5-Dihydroxy-1,4-benzoquinone
CAS 615-94-1
Molecular Formula C6H4O4
Quantity 1 g
Synonym 2,5-Dihydroxy-2,5-cyclohexadiene-1,4-dione, 2,5-Dihydroxyquinone, ,5-Dihydroxy-p-benzoquinone, 2,5-Dihydroxycyclohexa-2,5-diene-1,4-dione, Anilic Acid, NSC 3843
InChI Formula InChI=1S/C6H4O4/c7-3-1-4(8)6(10)2-5(3)9/h1-2,7,10H
SMILES OC1=CC(=O)C(=CC1=O)O
IUPAC Name 2,5-dihydroxycyclohexa-2,5-diene-1,4-dione
Molecular Weight (g/mol) 140.09
Formula Weight 140.011
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.