missing translation for 'onlineSavingsMsg'
Learn More

2,5-Dimethoxy-N-methylaniline, TRC

Product Code. 30375128
Change view
Click to view available options
Quantity:
25 mg
100 mg
250 mg
Unit Size:
250mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30375128 25 mg -
30372252 100 mg -
30386240 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30375128 Supplier Toronto Research Chemicals Supplier No. TRCD47280825MG
 Restricted Item
Estimated Shipment: 06-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10224-66-5
Molecular Formula C9H13NO2
Molecular Weight (g/mol) 167.21
Synonym N-Methyl-2,5-dimethoxyaniline, 2,5-Dimethoxy-N-methylbenzenamine, (2,5-Dimethoxy-phenyl)-methyl-amine
SMILES CNc1cc(OC)ccc1OC

Specifications

Quantity 25 mg
InChI Formula InChI=1S/C9H13NO2/c1-10-8-6-7(11-2)4-5-9(8)12-3/h4-6,10H,1-3H3
Formula Weight 167.0946
Chemical Name or Material 2,5-Dimethoxy-N-methylaniline
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.