missing translation for 'onlineSavingsMsg'
Learn More

2,5-Dimethyl-1,3,4-thiadiazole, 99+%, Thermo Scientific™

Product Code. 11421990
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11421990 25 g 25g
11411990 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11421990 Supplier Thermo Scientific Alfa Aesar Supplier No. A17227.14

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 27464-82-0
Molecular Formula C4H6N2S
Molecular Weight (g/mol) 114.166
MDL Number MFCD00020722
InChI Key JXQGICFGPUAVLJ-UHFFFAOYSA-N
PubChem CID 96922
IUPAC Name 2,5-dimethyl-1,3,4-thiadiazole
SMILES CC1=NN=C(S1)C

Specifications

Melting Point 61°C to 65°C
Quantity 25 g
Synonym 2,5-dimethylthiadiazole, 1,3,4-thiadiazole, 2,5-dimethyl, dimethyl-1,3,4-thiadiazole, unii-frm8gdw64z, frm8gdw64z, pubchem23972, acmc-1cr4f, 1,4-thiadiazole, 2,5-dimethyl, 1,3,4-thiadiazole,2,5-dimethyl, 2,5-dimethyl-1,3,4-thiadiazole
Formula Weight 114.17
Percent Purity ≥99%
Chemical Name or Material 2,5-Dimethyl-1,3,4-thiadiazole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.