missing translation for 'onlineSavingsMsg'
Learn More

2-[[[5-[(Dimethylamino)methyl]furan-2-yl]methyl]sulfanyl]ethanamine Hemifumarate, Mikromol™

Product Code. 30050551
Change view
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30050551 25 mg 25mg
30049159 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30050551 Supplier Mikromol Supplier No. MM0086.150025
 Restricted Item
Estimated Shipment: 08-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

2-[[[5-[(Dimethylamino)methyl]furan-2-yl]methyl]sulfanyl]ethanamine Hemifumarate
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2-[([5-[(Dimethylamino)methyl]furan-2-yl]methyl)sulfanyl]ethanamine hemifumarate
CAS 256948-32-0
Analyte or Component Names 2-[([5-[(Dimethylamino)methyl]furan-2-yl]methyl)sulfanyl]ethanamine hemifumarate
Shipping Condition Room Temperature
Molecular Formula 2 C10 H18 N2 O S . C4 H4 O4
Certifications/Compliance ISO 17025
Synonym 5-[[(2-aminoethyl)thio]methyl]-N,N-dimethyl-2-furanmethanamine (2E)-2-butenedioate (2:1), USP Ranitidine Related Compound A, AH 19013, Ph Eur Ranitidine Impurity B, BP Ranitidine Impurity B, USP Ranitidine Related Compound A, diamine (as AH 19013AB), Ranitidine Related Compound A as Hemifumarate, Ranitidine Hydrochloride Impurity B as Hemifumarate, Ranitidine Impurity B as Hemifumarate
InChI Formula InChI=1S/2C10H18N2OS.C4H4O4/c2*1-12(2)7-9-3-4-10(13-9)8-14-6-5-11;5-3(6)1-2-4(7)8/h2*3-4H,5-8,11H2,1-2H3;1-2H,(H,5,6)(H,7,8)/b;;2-1+
SMILES CN(C)Cc1oc(CSCCN)cc1.CN(C)Cc2oc(CSCCN)cc2.OC(=O)\C=C\C(=O)O
IUPAC Name (E)-but-2-enedioic acid;2-[[5-[(dimethylamino)methyl]furan-2-yl]methylsulfanyl]ethanamine
Molecular Weight (g/mol) 544.7276
Quantity 25 mg
Formula Weight 544.2389 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.