missing translation for 'onlineSavingsMsg'
Learn More

2,5-Dimethylfuran, 98+%

Product Code. 11434070
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11434070 500 g 500g
11405233 25 g 25g
11415233 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11434070 Supplier Thermo Scientific Alfa Aesar Supplier No. A12833.36
 Restricted Item
Estimated Shipment: 25-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 625-86-5 | C6H8O | 96.129 g/mol

2,5-Dimethylfuran is a major metabolite of hexane with singlet oxygen scavenging ability. It is used as biofuel, biomaker for smoking, pharmaceutical intermediate and in organic synthesis. It is also used to analyze samples for the derivatization of carbonyl-containing compounds to their oximes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,5-Dimethylfuran is a major metabolite of hexane with singlet oxygen scavenging ability. It is used as biofuel, biomaker for smoking, pharmaceutical intermediate and in organic synthesis. It is also used to analyze samples for the derivatization of carbonyl-containing compounds to their oximes.

Solubility
Slightly miscible with water. Miscible with ethanol and fats.

Notes
Incompatible with strong oxidizing agents, strong acids and strong bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 625-86-5
Molecular Formula C6H8O
Molecular Weight (g/mol) 96.129
MDL Number MFCD00003250
InChI Key GSNUFIFRDBKVIE-UHFFFAOYSA-N
Synonym furan, 2,5-dimethyl, 2,5-dimethylfurane, 2,5-dimethyl-furan, 2,5-dimethyl furane, unii-dr5hl9oj7y, ccris 3158, 2,5-dimethyl furan, dr5hl9oj7y, 2.5-dimethyl furan, acmc-1avpg
PubChem CID 12266
IUPAC Name 2,5-dimethylfuran
SMILES CC1=CC=C(O1)C

Specifications

Melting Point -62°C
Density 0.903
Boiling Point 92°C to 93°C
Flash Point 1°C (33°F)
Refractive Index 1.441
Quantity 500 g
UN Number UN3271
Beilstein 106449
Solubility Information Slightly miscible with water. Miscible with ethanol and fats.
Formula Weight 96.13
Percent Purity ≥98%
Physical Form Liquid
Chemical Name or Material 2,5-Dimethylfuran
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 2,5-Dimethylfuran
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 2
  • Skin sensitiser Category 1
  • Specific target organ toxicity Category 3
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H317-May cause an allergic skin reaction.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
Supplemental information
  • MIXTURE LIST-Contain: 2,5-Dimethylfuran

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.