missing translation for 'onlineSavingsMsg'
Learn More

2,5-Dimethylfuran, TRC

Product Code. 30137139
Change view
Click to view available options
Quantity:
1 g
10 g
100 g
Unit Size:
100g
10g
1g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30137139 10 g 10g
30009509 100 g 100g
17952265 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30137139 Supplier Toronto Research Chemicals Supplier No. TRCD47305010G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 625-86-5
Molecular Formula C6 H8 O
Molecular Weight (g/mol) 96.13
Synonym 2,5-Dimethylfuran, 2-Methyl-5-methylfuran, NSC 6220
IUPAC Name 2,5-dimethylfuran
SMILES Cc1oc(C)cc1

Specifications

Quantity 10 g
InChI Formula InChI=1S/C6H8O/c1-5-3-4-6(2)7-5/h3-4H,1-2H3
Formula Weight 96.0575
Chemical Name or Material 2,5-Dimethylfuran
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.