missing translation for 'onlineSavingsMsg'
Learn More

Hydroxy Brimonidine Trifluoroacetic Acid Salt, TRC

Product Code. 17952605
Change view
Click to view available options
Quantity:
2.5 mg
25 mg
Unit Size:
2.5mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17952605 2.5 mg 2.5mg
17982605 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17952605 Supplier Toronto Research Chemicals Supplier No. TRCH8300002.5MG
 Restricted Item
Estimated Shipment: 14-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1391054-10-6
Molecular Formula C11 H12 Br N5 O . C2 H F3 O2
Molecular Weight (g/mol) 424.1732
Synonym Brimonidine EP Impurity G
IUPAC Name 1-(2-aminoethyl)-3-(5-bromoquinoxalin-6-yl)urea;2,2,2-trifluoroacetic acid
SMILES NCCNC(=O)Nc1ccc2nccnc2c1Br.OC(=O)C(F)(F)F

Specifications

Quantity 2.5 mg
InChI Formula InChI=1S/C11H12BrN5O.C2HF3O2/c12-9-7(17-11(18)16-4-3-13)1-2-8-10(9)15-6-5-14-8;3-2(4,5)1(6)7/h1-2,5-6H,3-4,13H2,(H2,16,17,18);(H,6,7)
Formula Weight 423.0154
Percent Purity >95%
Chemical Name or Material Hydroxy Brimonidine Trifluoroacetic Acid Salt
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.