Learn More
2-[6-(Acetylphenylamino)-1,3,5-hexatrienyl]-1,1-dimethyl-3-sulfobutyl-1H-benz[e]indolium Inner Salt, TRC
Product Code. 30010533
Change view
Click to view available options
Quantity:
100 mg
1 g
Unit Size:
100mg
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30010533 100 mg 100mg
30005161 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 30010533 Supplier Toronto Research Chemicals Supplier No. TRCA187940100MG
 Restricted Item
Estimated Shipment: 28-08-2026
Add to basket
Add to basket
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 63450-66-8
Molecular Formula C32 H34 N2 O4 S
Molecular Weight (g/mol) 542.69
IUPAC Name 4-[2-[(1E,3E,5E)-6-(N-acetylanilino)hexa-1,3,5-trienyl]-1,1-dimethyl-benzo[e]indol-3-ium-3-yl]butane-1-sulfonate
SMILES CC(=O)N(\C=C\C=C\C=C\C1=[N+](CCCCS(=O)(=O)[O-])c2ccc3ccccc3c2C1(C)C)c4ccccc4
Specifications
Quantity 100 mg
InChI Formula InChI=1S/C32H34N2O4S/c1-25(35)33(27-16-7-6-8-17-27)22-12-5-4-9-19-30-32(2,3)31-28-18-11-10-15-26(28)20-21-29(31)34(30)23-13-14-24-39(36,37)38/h4-12,15-22H,13-14,23-24H2,1-3H3
Formula Weight 542.2239
Chemical Name or Material 2-[6-(Acetylphenylamino)-1,3,5-hexatrienyl]-1,1-dimethyl-3-sulfobutyl-1H-benz[e]indolium Inner Salt