Learn More
2',6-Dibromo-3,4'-bipyridine, 95%, Thermo Scientific™
Product Code. 11352199
Change view
Click to view available options
Quantity:
100 mg
500 mg
Unit Size:
100mg
500mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11352199 500 mg 500mg
11342199 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11352199 Supplier Thermo Scientific Alfa Aesar Supplier No. H27214.ME
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
missing translation for 'validMainframeMsg'
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 942206-16-8
Molecular Formula C10H6Br2N2
Molecular Weight (g/mol) 313.98
MDL Number MFCD09037424
InChI Key LGVJILYMUFZGLC-UHFFFAOYSA-N
Synonym 2',6-dibromo-3,4'-bipyridine, 6,2'-dibromo-3,4'-bipyridine, 2-bromo-5-2-bromopyridin-4-yl pyridine, 6,2'-dibromo-3,4' bipyridine
PubChem CID 42553011
IUPAC Name 2-bromo-4-(6-bromopyridin-3-yl)pyridine
SMILES BrC1=NC=C(C=C1)C1=CC(Br)=NC=C1
Especificaciones
Melting Point 189°C to 193°C
Quantity 500 mg
Formula Weight 313.98
Percent Purity 95%
Chemical Name or Material 2',6-Dibromo-3,4'-bipyridine
Título del producto
Seleccione un problema
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.