Learn More
2,6-Dichloro-4-(trifluoromethyl)pyridine, 98%, Thermo Scientific™
Product Code. 15433557
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15433557 250 mg 250mg
11337059 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15433557 Supplier Thermo Scientific Alfa Aesar Supplier No. H26873.MD
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
missing translation for 'validMainframeMsg'
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 39890-98-7
Molecular Formula C6H2Cl2F3N
Molecular Weight (g/mol) 215.98
MDL Number MFCD00042246
InChI Key KVNQWVYYVLCZKK-UHFFFAOYSA-N
Synonym 2,6-dichloro-4-trifluoromethyl pyridine, 2,6-dichloro-4-trifluoromethyl-pyridine, 4-trifluoromethyl-2,6-dichloropyridine, 2,6-dichloro-4-trifluoromethylpyridine, acmc-1air6, abbypharma ap-30-1899, pyridine, 2,6-dichloro-4-trifluoromethyl
PubChem CID 520968
IUPAC Name 2,6-dichloro-4-(trifluoromethyl)pyridine
SMILES FC(F)(F)C1=CC(Cl)=NC(Cl)=C1
Specifications
Boiling Point 70°C to 75°C (25 mmHg)
Quantity 250 mg
UN Number UN2810
Formula Weight 215.99
Percent Purity 98%
Chemical Name or Material 2,6-Dichloro-4-(trifluoromethyl)pyridine