Learn More
2,6-Dichloroaniline-3,4,5-d3, TRC
Specifications
Specifications
| Chemical Name or Material | 2,6-Dichloroaniline-3,4,5-d3 |
| CAS | 77435-48-4 |
| Molecular Formula | C6 D3 H2 Cl2 N |
| Quantity | 10 mg |
| Synonym | Benzen-3,4,5-d3-amine, 2,6-dichloro- (9CI), 2,6-Dichlorobenzen-3,4,5-d3-amine, 2,6-Dichloroaniline-3,4,5-d3, 2,6-Dichloroaniline-D3 |
| InChI Formula | InChI=1S/C6H5Cl2N/c7-4-2-1-3-5(8)6(4)9/h1-3H,9H2/i1D,2D,3D |
| SMILES | [2H]c1c([2H])c(Cl)c(N)c(Cl)c1[2H] |
| IUPAC Name | 2,6-dichloro-3,4,5-trideuterioaniline |
| Molecular Weight (g/mol) | 165.0351 |
| Formula Weight | 163.9987 |