Learn More
2,6-Dichloroformanilide, TRC
Product Code. 30387798
Change view
Click to view available options
Quantity:
1 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30387798 10 g
30370656 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 30387798 Supplier Toronto Research Chemicals Supplier No. TRCD43440010G
 Restricted Item
This item requires a price update prior to being ordered.
Request a Price for this item here.
missing translation for 'validMainframeMsg'
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 10113-35-6
Molecular Formula C7H5Cl2NO
Molecular Weight (g/mol) 190.03
Synonym 2,6-Dichloro-N-formylaniline, 2,6-Dichloroformanilide, 2,6-Dichlorophenylformamide, 2',6'-Dichloroformanilide, N-(2,6-Dichlorophenyl)formamide
SMILES Clc1cccc(Cl)c1NC=O
Specifications
Quantity 10 g
InChI Formula InChI=1S/C7H5Cl2NO/c8-5-2-1-3-6(9)7(5)10-4-11/h1-4H,(H,10,11)
Formula Weight 188.9748
Chemical Name or Material 2,6-Dichloroformanilide