missing translation for 'onlineSavingsMsg'
Learn More

2,6-Dichlorophenylacetic Acid Methyl Ester, TRC

Product Code. 30380969
Change view
Click to view available options
Quantity:
2.5 g
10 g
Unit Size:
10g
2.5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30380969 2.5 g 2.5g
30386167 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30380969 Supplier Toronto Research Chemicals Supplier No. TRCD4356552.5G
 Restricted Item
Estimated Shipment: 30-07-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2,6-Dichlorophenylacetic Acid Methyl Ester
CAS 54551-83-6
Molecular Formula C9 H8 Cl2 O2
Quantity 2.5 g
Synonym 2,6-Dichloro-benzeneacetic Acid Methyl Ester, Methyl (2,6-Dichlorophenyl)acetate
InChI Formula InChI=1S/C9H8Cl2O2/c1-13-9(12)5-6-7(10)3-2-4-8(6)11/h2-4H,5H2,1H3
SMILES COC(=O)Cc1c(Cl)cccc1Cl
IUPAC Name methyl 2-(2,6-dichlorophenyl)acetate
Molecular Weight (g/mol) 219.06
Formula Weight 217.9901
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.