Learn More
2,6-Difluoroanisole, 98%, Thermo Scientific™
Product Code. 15545246 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15545246 1 g 1g
15555246 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15545246 Supplier Thermo Scientific Acros Supplier No. 457450010
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Chemical Identifiers
CAS 437-82-1
Molecular Formula C7H6F2O
Molecular Weight (g/mol) 144.12
InChI Key IOBWAHRFIPQEQL-UHFFFAOYSA-N
Synonym 2,6-difluoroanisole, 1,3-difluoro-2-methoxy-benzene, benzene, 1,3-difluoro-2-methoxy, pubchem1968, 2,6-difluoro anisole, 2.6-difluoro anisole, acmc-1ajjj, ksc235o3h, iobwahrfipqeql-uhfffaoysa
PubChem CID 2736897
IUPAC Name 1,3-difluoro-2-methoxybenzene
SMILES COC1=C(C=CC=C1F)F
Specifications
Density 1.2210g/mL
Boiling Point 70.0°C to 72.0°C (56.0 mmHg)
Flash Point 42°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass Bottle
Quantity 1 g
Specific Gravity 1.221
Formula Weight 144.12
Percent Purity 98%
Chemical Name or Material 2,6-Difluoroanisole
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 2,6-Difluoroanisole, 98%
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.