missing translation for 'onlineSavingsMsg'
Learn More

2,6-Difluoroanisole, 98%, Thermo Scientific™

Product Code. 15555246 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15555246 5 g 5g
15545246 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15555246 Supplier Thermo Scientific Acros Supplier No. 457450050

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 437-82-1
Molecular Formula C7H6F2O
Molecular Weight (g/mol) 144.12
InChI Key IOBWAHRFIPQEQL-UHFFFAOYSA-N
Synonym 2,6-difluoroanisole, 1,3-difluoro-2-methoxy-benzene, benzene, 1,3-difluoro-2-methoxy, pubchem1968, 2,6-difluoro anisole, 2.6-difluoro anisole, acmc-1ajjj, ksc235o3h, iobwahrfipqeql-uhfffaoysa
PubChem CID 2736897
IUPAC Name 1,3-difluoro-2-methoxybenzene
SMILES COC1=C(C=CC=C1F)F

Specifications

Density 1.2210g/mL
Boiling Point 70.0°C to 72.0°C (56.0 mmHg)
Flash Point 42°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Quantity 5 g
Specific Gravity 1.221
Formula Weight 144.12
Percent Purity 98%
Chemical Name or Material 2,6-Difluoroanisole
compliance-icons compliance-icons
Product Identifier
  • 2,6-Difluoroanisole, 98%
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.