missing translation for 'onlineSavingsMsg'
Learn More

2,6-Difluorophenyl isocyanate, 97%, ACROS Organics™

Product Code. 10175700 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10175700 1 g Glass bottle 1g
10289263 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 10175700 Supplier Thermo Scientific Acros Supplier No. 308150010

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Chemical Identifiers

CAS 65295-69-4
Molecular Formula C7H3F2NO
Molecular Weight (g/mol) 155.104
MDL Number MFCD00001998
InChI Key YXHDLKWTPVMIOH-UHFFFAOYSA-N
Synonym 2,6-difluorophenyl isocyanate, 2,6-difluorophenylisocyanate, benzene, 1,3-difluoro-2-isocyanato, 2,6-difluorobenzenisocyanate, timtec-bb sbb006664, pubchem5023, acmc-1bcwu, 2,6 difluorophenylisocyanate, 2,6-difluoro phenyl isocyanate, 1-isocyanato-2,6-difluoro benzene
PubChem CID 522162
IUPAC Name 1,3-difluoro-2-isocyanatobenzene
SMILES C1=CC(=C(C(=C1)F)N=C=O)F

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 22.0°C to 25.0°C
Color White to Yellow
Flash Point 58°C
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Linear Formula F2C6H3NCO
Refractive Index 1.4945 to 1.4965
Quantity 1 g
Formula Weight 155.1
Percent Purity 97%
Physical Form Liquid or Low Melting Solid
Chemical Name or Material 2, 6-Difluorophenyl isocyanate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.