missing translation for 'onlineSavingsMsg'
Learn More

2,6-Difluorophenyl isocyanate, 97%

Product Code. 11303686
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11303686 1 g 1g
11313686 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11303686 Supplier Thermo Scientific Alfa Aesar Supplier No. L14779.03

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 65295-69-4 | C7H3F2NO | 155.104 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 65295-69-4
Molecular Formula C7H3F2NO
Molecular Weight (g/mol) 155.104
MDL Number MFCD00001998
InChI Key YXHDLKWTPVMIOH-UHFFFAOYSA-N
Synonym 2,6-difluorophenyl isocyanate, 2,6-difluorophenylisocyanate, benzene, 1,3-difluoro-2-isocyanato, 2,6-difluorobenzenisocyanate, timtec-bb sbb006664, pubchem5023, acmc-1bcwu, 2,6 difluorophenylisocyanate, 2,6-difluoro phenyl isocyanate, 1-isocyanato-2,6-difluoro benzene
PubChem CID 522162
IUPAC Name 1,3-difluoro-2-isocyanatobenzene
SMILES C1=CC(=C(C(=C1)F)N=C=O)F

Specifications

Melting Point 22°C to 25°C
Boiling Point 60°C (10 mmHg)
Flash Point 58°C (136°F)
Refractive Index 1.4928
Quantity 1 g
UN Number UN2930
Beilstein 7115873
Sensitivity Moisture sensitive
Formula Weight 155.11
Percent Purity 97%
Chemical Name or Material 2,6-Difluorophenyl isocyanate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.