missing translation for 'onlineSavingsMsg'
Learn More

2-(6-Methoxy-2-naphthyl)acetonitrile, Mikromol

Product Code. 30460040
Change view
Click to view available options
Quantity:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30460040 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30460040 Supplier Mikromol Supplier No. MM0152.280025
 Restricted Item
Estimated Shipment: 25-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Discover Mikromol- your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

2-(6-Methoxy-2-naphthyl)acetonitrile, Mikromol
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2-(6-Methoxy-2-naphthyl)acetonitrile
CAS 71056-96-7
Analyte or Component Names 2-(6-Methoxy-2-naphthyl)acetonitrile
Shipping Condition Room Temperature
Molecular Formula C13 H11 N O
Certifications/Compliance ISO/IEC 17025
InChI Formula InChI=1S/C13H11NO/c1-15-13-5-4-11-8-10(6-7-14)2-3-12(11)9-13/h2-5,8-9H,6H2,1H3
Solution Type Neat
SMILES COc1ccc2cc(CC#N)ccc2c1
IUPAC Name 2-(6-methoxynaphthalen-2-yl)acetonitrile
Molecular Weight (g/mol) 197.23
Quantity 25 mg
Formula Weight 197.0841 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.