missing translation for 'onlineSavingsMsg'
Learn More

2-(6-Methoxy-2-naphthyl)acetonitrile, TRC

Product Code. 30482756
Change view
Click to view available options
Quantity:
50 mg
100 mg
250 mg
Unit Size:
100mg
250mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30482756 250 mg 250mg
30480808 50 mg 50mg
30468169 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30482756 Supplier Toronto Research Chemicals Supplier No. TRCM264730250MG
 Restricted Item
Estimated Shipment: 04-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 71056-96-7
Molecular Formula C13 H11 N O
Molecular Weight (g/mol) 197.23
Synonym 2-Naphthaleneacetonitrile, 6-methoxy-, 6-Methoxy-2-naphthaleneacetonitrile, 2-(6-Methoxy-2-naphthyl)acetonitrile, 2-(6-Methoxy-2-naphthyl)ethanenitrile, 2-Cyanomethyl-6-methoxynaphthalene, 6-Methoxynaphthalen-2-ylacetonitrile
IUPAC Name 2-(6-methoxynaphthalen-2-yl)acetonitrile
SMILES COc1ccc2cc(CC#N)ccc2c1

Specifications

Quantity 250 mg
InChI Formula InChI=1S/C13H11NO/c1-15-13-5-4-11-8-10(6-7-14)2-3-12(11)9-13/h2-5,8-9H,6H2,1H3
Formula Weight 197.0841
Chemical Name or Material 2-(6-Methoxy-2-naphthyl)acetonitrile
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.