missing translation for 'onlineSavingsMsg'
Learn More

2,7-Dibromo-9,9-dimethylfluorene, 98%, Thermo Scientific™

Product Code. 15535956 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15535956 5 g 5g
15545956 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15535956 Supplier Thermo Scientific Acros Supplier No. 457310050

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 28320-32-3
Molecular Formula C15H12Br2
Molecular Weight (g/mol) 352.07
MDL Number MFCD00099471
InChI Key LONBOJIXBFUBKQ-UHFFFAOYSA-N
Synonym 2,7-dibromo-9,9-dimethyl-9h-fluorene, 9,9-dimethyl-2,7-dibromofluorene, 2,7-dibromo-9,9-dimethyl-fluorene, 9h-fluorene, 2,7-dibromo-9,9-dimethyl, 2,7-dibromo-9,9-dimethyl fluorene, pubchem19656, acmc-1cpi8, ksc491i1n, 9,9-dimethyl-2,7-dibromofluorenone
PubChem CID 12123876
IUPAC Name 2,7-dibromo-9,9-dimethylfluorene
SMILES CC1(C)C2=C(C=CC(Br)=C2)C2=C1C=C(Br)C=C2

Specifications

Melting Point 179.0°C to 181.0°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Quantity 5 g
Formula Weight 352.07
Percent Purity 98%
Chemical Name or Material 2,7-Dibromo-9,9-dimethylfluorene
compliance-icons
Product Identifier
  • 2,7-Dibromo-9,9-dimethylfluorene, 98%
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.