missing translation for 'onlineSavingsMsg'
Learn More

2,7-Dichlorofluorene, TRC

Product Code. 30374865
Change view
Click to view available options
Quantity:
1 g
2.5 g
Unit Size:
1g
2.5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30374865 1 g 1g
30380124 2.5 g 2.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30374865 Supplier Toronto Research Chemicals Supplier No. TRCD4344651G
 Restricted Item
Estimated Shipment: 03-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7012-16-0
Molecular Formula C13 H8 Cl2
Molecular Weight (g/mol) 235.11
Synonym 2,7-Dichloro-9H-fluorene, NSC 73077
IUPAC Name 2,7-dichloro-9H-fluorene
SMILES Clc1ccc2c(Cc3cc(Cl)ccc23)c1

Specifications

Quantity 1 g
InChI Formula InChI=1S/C13H8Cl2/c14-10-1-3-12-8(6-10)5-9-7-11(15)2-4-13(9)12/h1-4,6-7H,5H2
Formula Weight 234.0003
Chemical Name or Material 2,7-Dichlorofluorene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.