missing translation for 'onlineSavingsMsg'
Learn More

2,8-Bis-(trifluoromethyl)-4-quinolinol, Mikromol™

Product Code. 30050858
Change view
Click to view available options
Quantity:
25 mg
100 mg
4 x 25 mg
Unit Size:
100mg
25mg
Product Code. Quantity unitSize
30050858 100 mg 100mg
30052077 25 mg 25mg
30459478 4 x 25 mg -
3 options
This item is not returnable. View return policy

Product Code. 30050858

Brand: Mikromol MM1198.01

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

2,8-Bis-(trifluoromethyl)-4-quinolinol
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2,8-Bis-(trifluoromethyl)-4-quinolinol
CAS 35853-41-9
Analyte or Component Names 2,8-Bis-(trifluoromethyl)-4-quinolinol
Shipping Condition Room Temperature
Molecular Formula C11 H5 F6 N O
Certifications/Compliance ISO 17025
Synonym 2,8-Bis-(trifluoromethyl)-4-quinolinol
InChI Formula InChI=1S/C11H5F6NO/c12-10(13,14)6-3-1-2-5-7(19)4-8(11(15,16)17)18-9(5)6/h1-4H,(H,18,19)
SMILES Oc1cc(nc2c(cccc12)C(F)(F)F)C(F)(F)F
IUPAC Name 2,8-bis(trifluoromethyl)quinolin-4-ol
Molecular Weight (g/mol) 281.15
Quantity 100 mg
Formula Weight 281.0275 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.