missing translation for 'onlineSavingsMsg'
Learn More

2,8-Dimethylimidazo[1,2-a]pyridine-3-carboxylic acid, 95%, Thermo Scientific™

Product Code. 11419350
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
This item is not returnable. View return policy

Product Code. 11419350

Brand: Thermo Scientific Alfa Aesar H50233.MD

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 874605-59-1
Molecular Formula C10H10N2O2
Molecular Weight (g/mol) 190.202
MDL Number MFCD00269632
InChI Key CAFXSRDUIAZBHO-UHFFFAOYSA-N
Synonym 2,8-dimethylimidazo 1,2-a pyridine-3-carboxylic acid, 2,8-dimethylimidazo 1,2-a pyridine-3-carboxylicacid, 2,8-dimethyl-imidazo 1,2-a pyridine-3-carboxylic acid, imidazo 1,2-a pyridine-3-carboxylic acid, 2,8-dimethyl, acmc-20ao79, dimethylimidazoapyridinecarboxylicacid, imidazo 1,2-a pyridine-3-carboxylicacid, 2,8-dimethyl, 2,8-dimethyl-4-hydroimidazo 1,2-a pyridine-3-carboxylic acid
PubChem CID 2736259
IUPAC Name 2,8-dimethylimidazo[1,2-a]pyridine-3-carboxylic acid
SMILES CC1=CC=CN2C1=NC(=C2C(=O)O)C

Specifications

Quantity 250 mg
Formula Weight 190.2
Percent Purity 95%
Chemical Name or Material 2,8-Dimethylimidazo[1,2-a]pyridine-3-carboxylic acid
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.