Learn More
2-(Acetyloxy)-3-methylbenzoic acid, 97%, Thermo Scientific™
Product Code. 10633223
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
Unit Size:
10g
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10633223 10 g 10g
10712693 1 g 1g
10269924 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 10633223 Supplier Thermo Scientific Maybridge Supplier No. SB01734EA
 Restricted Item
Estimated Shipment: 06-10-2026
Add to basket
missing translation for 'validMainframeMsg'
Chemical Identifiers
CAS 4386-39-4
Molecular Formula C10H10O4
Molecular Weight (g/mol) 194.19
MDL Number MFCD00085683
InChI Key XRBMKGUDDJPAMH-UHFFFAOYSA-N
Synonym 2-acetyloxy-3-methylbenzoic acid, 2-acetoxy-3-methylbenzoic acid, 2-acetoxy-m-toluic acid, unii-2g83e641hz, 2-acetoxy-3-methyl-benzoic acid, 3-methylacetylsalicylic acid, benzoic acid,2-acetyloxy-3-methyl, 3-methyl-2-acetoxybenzoic acid, cresopirine, cresopyrine
PubChem CID 78094
IUPAC Name 2-acetyloxy-3-methylbenzoic acid
SMILES CC(=O)OC1=C(C)C=CC=C1C(O)=O
Specifications
CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber glass bottle
Quantity 10 g
Formula Weight 194.19
Percent Purity 97%
Chemical Name or Material 2-(Acetyloxy)-3-methylbenzoic acid