Learn More
2-Acrylamide-2-methylpropanesulfonic Acid, TRC
Product Code. 30328412
Change view
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30328412 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30328412 Supplier Toronto Research Chemicals Supplier No. TRCA191328500MG
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 2-Acrylamido-2-methylpropanesulphonic acid
CAS 15214-89-8
Molecular Formula C7 H13 N O4 S
Quantity 500 mg
Synonym 1-Propanesulfonic acid, 2-acrylamido-2-methyl- (8CI), 1-Propanesulfonic acid, 2-methyl-2-[(1-oxo-2-propenyl)amino]- (9CI), 2-Methyl-2-[(1-oxo-2-propen-1-yl)amino]-1-propanesulfonic acid, 2-Acrylamido-2,2-dimethylethanesulfonic acid, 2-Acrylamido-2-methyl-1-propanesulfonic acid, 2-Acrylamido-2-methylpropanesulfonic acid, 2-Acrylamido-2-methylpropylsulfonic acid, 2-Acryloylamido-2-methylpropanesulfonic acid, 2-Acryloylamino-2-methyl-1-propanesulfonic acid, 2-Methyl-2-[(1-oxo-2-propenyl)amino]-1-propanesulfonic acid, AMPS, AMPS (sulfonic acid), ATBS, Acrylamide tert-butylsulfonic acid, CG 810S-P, Lubrizol 2402, Lubrizol 2404, Lubrizol AMPS, TBAS-Q, tert-Butylacrylamidosulfonic acid
InChI Formula InChI=1S/C7H13NO4S/c1-4-6(9)8-7(2,3)5-13(10,11)12/h4H,1,5H2,2-3H3,(H,8,9)(H,10,11,12)
SMILES CC(C)(CS(=O)(=O)O)NC(=O)C=C
IUPAC Name 2-methyl-2-(prop-2-enoylamino)propane-1-sulfonic acid
Molecular Weight (g/mol) 207.247
Formula Weight 207.0565