missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-1,6-dimethylfuro[3,2-e]imidazo[4,5-b]pyridine, TRC

Product Code. 17970555
Change view
Click to view available options
Quantity:
0.5 mg
5 mg
Unit Size:
0.5mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17970555 5 mg 5mg
17964745 0.5 mg 0.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17970555 Supplier Toronto Research Chemicals Supplier No. TRCA6050705MG
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 357383-27-8
Molecular Formula C10 H10 N4 O
Molecular Weight (g/mol) 202.2126
Synonym 1,6-Dimethyl-1H-furo[3,2-b]imidazo[4,5-e]pyridin-2-amine, IFP
SMILES CN1C(=N)Nc2nc3cc(C)oc3cc12

Specifications

Quantity 5 mg
InChI Formula InChI=1S/C10H10N4O/c1-5-3-6-8(15-5)4-7-9(12-6)13-10(11)14(7)2/h3-4H,1-2H3,(H2,11,12,13)
Formula Weight 202.0855
Chemical Name or Material 2-Amino-1,6-dimethylfuro[3,2-e]imidazo[4,5-b]pyridine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.