missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-4,6-dichloropyrimidine-5-carboxaldehyde, 96%, Thermo Scientific Chemicals

Product Code. 15457735
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 15457735

Brand: Thermo Scientific Alfa Aesar H27844.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5604-46-6
Molecular Formula C5H3Cl2N3O
Molecular Weight (g/mol) 192.00
MDL Number MFCD03001242
InChI Key GOJUJUVQIVIZAV-UHFFFAOYSA-N
Synonym 2-amino-4,6-dichloro-5-formylpyrimidine, 2-amino-4,6-dichloro-5-pyrimidinecarbaldehyde, 2-amino-4,6-dichloro-5-formyl-pyrimidine, 2-amino-4.6-dichloropyrimidine-5-carbaldehyde, 2-amino-4,6-dichloro-pyrimidine-5-carbaldehyde, 2-amino-4,6-dichloropyrimidine-5-carboxaldehyde, 5-pyrimidinecarboxaldehyde, 2-amino-4,6-dichloro, 2-amino, pubchem6979, acmc-209lqn
PubChem CID 265546
IUPAC Name 2-amino-4,6-dichloropyrimidine-5-carbaldehyde
SMILES NC1=NC(Cl)=C(C=O)C(Cl)=N1

Specifications

Melting Point ∼220°C
Quantity 5 g
Sensitivity Air Sensitive
Formula Weight 192
Percent Purity 96%
Chemical Name or Material 2-Amino-4,6-dichloropyrimidine-5-carboxaldehyde

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.