missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Amino-4-chloro-6-methoxy-pyrimidine, TRC
Click to view available options
Quantity:
50 g
Unit Size:
50g
Specifications
Specifications
| Chemical Name or Material | 2-Amino-4-chloro-6-methoxy-pyrimidine |
| CAS | 5734-64-5 |
| Molecular Formula | C5H6ClN3O |
| Quantity | 50 g |
| Synonym | 4-Chloro-6-methoxy-2-pyrimidinamine, (4-Chloro-6-methoxypyrimidin-2-yl)amine, 2-Amino-6-chloro-4-methoxypyrimidine, 4-Chloro-6-methoxy-2-pyrimidinamine, 4-Methoxy-6-chloro-2-aminopyrimidine, NSC 28420 |
| InChI Formula | InChI=1S/C5H6ClN3O/c1-10-4-2-3(6)8-5(7)9-4/h2H,1H3,(H2,7,8,9) |
| SMILES | COc1cc(Cl)nc(N)n1 |
| IUPAC Name | 4-chloro-6-methoxypyrimidin-2-amine |
| Molecular Weight (g/mol) | 159.57 |
| Formula Weight | 159.0199 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction