missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-4-chlorophenol, TRC

Product Code. 17944345
Change view
Click to view available options
Quantity:
5 g
50 g
100 g
Unit Size:
100g
50g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17944345 50 g 50g
17914095 100 g 100g
17914815 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17944345 Supplier Toronto Research Chemicals Supplier No. TRCA60327550G

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 95-85-2
Molecular Formula C6 H6 Cl N O
Molecular Weight (g/mol) 143.57
Synonym 2-Amino-4-chlorophenol, 1-Amino-2-hydroxy-5-chlorobenzene, 1-Hydroxy-2-amino-4-chlorobenzene, 2-Hydroxy-5-chloroaniline, 4-Chloro-2-aminophenol, 4-Chloro-o-aminophenol, 5-Chloro-2-hydroxyaniline, C.I. 76525, C.I. Oxidation Base 18, Fouramine PY, NSC 247814, o-Amino-p-chlorophenol, p-Chloro-o-aminophenol
IUPAC Name 2-amino-4-chlorophenol
SMILES Nc1cc(Cl)ccc1O

Specifications

Quantity 50 g
InChI Formula InChI=1S/C6H6ClNO/c7-4-1-2-6(9)5(8)3-4/h1-3,9H,8H2
Formula Weight 143.0138
Chemical Name or Material 2-Amino-4-chlorophenol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.