Learn More
2-Amino-4-methoxypyridine, TRC
Product Code. 30323597
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30323597 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30323597 Supplier Toronto Research Chemicals Supplier No. TRCA6119951G
 Restricted Item
Estimated Shipment: 12-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 2-Amino-4-methoxypyridine
CAS 10201-73-7
Molecular Formula C6H8N2O
Quantity 1 g
Synonym (4-Methoxypyridin-2-yl)amine, 4-(Methyloxy)-2-pyridinamine, 4-Methoxy-2-aminopyridine, 4-Methoxypyridin-2-amine
InChI Formula InChI=1S/C6H8N2O/c1-9-5-2-3-8-6(7)4-5/h2-4H,1H3,(H2,7,8)
SMILES COc1ccnc(N)c1
IUPAC Name 4-methoxypyridin-2-amine
Molecular Weight (g/mol) 124.14
Formula Weight 124.0637