Learn More
2-Amino-5-(4-(2-(5-ethylpyridin-2-yl)ethoxy)benzyl)thiazol-4(5H)-one, 95%, BLD Pharm™
Specifications
Specifications
| CAS | 105355-26-8 |
| Molecular Formula | C19H21N3O2S |
| MDL Number | MFCD07368627 |
| Quantity | 1 g |
| SMILES | O=C1N=C(N)SC1CC2=CC=C(OCCC3=NC=C(CC)C=C3)C=C2 |
| Molecular Weight (g/mol) | 355.45 |
| Percent Purity | 95% |
| Chemical Name or Material | 2-Amino-5-(4-(2-(5-ethylpyridin-2-yl)ethoxy)benzyl)thiazol-4(5H)-one |