missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-5-chloropyrimidine, 97%, Thermo Scientific™

Product Code. 11354126
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11354126 250 mg 250mg
11334126 1 g 1g
11344126 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11354126 Supplier Thermo Scientific Alfa Aesar Supplier No. L19248.MD

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5428-89-7
Molecular Formula C4H4ClN3
Molecular Weight (g/mol) 129.547
MDL Number MFCD00014604
InChI Key OQZMDDKDHRIGDY-UHFFFAOYSA-N
Synonym 2-amino-5-chloropyrimidine, 2-pyrimidinamine, 5-chloro, 5-chloro-2-pyrimidinamine, pyrimidine, 2-amino-5-chloro, 5-chloropyrimidine-2-ylamine, 5-chlorpyrimidin-2-amin, chloropyrimidinamine, zlchem 634, pubchem9619, acmc-1aya8
PubChem CID 79479
IUPAC Name 5-chloropyrimidin-2-amine
SMILES C1=C(C=NC(=N1)N)Cl

Specifications

Melting Point 235°C
Quantity 250 mg
Beilstein 110914
Formula Weight 129.55
Percent Purity 97%
Chemical Name or Material 2-Amino-5-chloropyrimidine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.