missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Amino-6-methylpyridine, TRC
missing translation for 'orderingAttributeHoverText'
Quantity:
50 g
missing translation for 'unitSize'
50g
Specifications
Specifications
| Chemical Name or Material | 2-Amino-6-methylpyridine |
| CAS | 1824-81-3 |
| Molecular Formula | C6H8N2 |
| Quantity | 50 g |
| Synonym | 6-Methyl-2-pyridinamine, 2-Amino-6-picoline, 6-Amino-2-methylpyridine, 6-Amino-2-picoline, 6-Methyl-2-aminopyridine, 6-Methyl-2-pyridylamine, 6-Methylpyridin-2-amine, 6-Methylpyridin-2-ylamine, NSC 1488, NSC 176163, NSC 6971 |
| InChI Formula | InChI=1S/C6H8N2/c1-5-3-2-4-6(7)8-5/h2-4H,1H3,(H2,7,8) |
| SMILES | Cc1cccc(N)n1 |
| IUPAC Name | 6-methylpyridin-2-amine |
| Molecular Weight (g/mol) | 108.14 |
| Formula Weight | 108.0687 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction