missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-6-nitrobenzothiazole, 96%

Product Code. 10133971 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
Packaging:
Plastic bottle
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10133971 25 g Plastic bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10133971 Supplier Thermo Scientific Acros Supplier No. 183740250

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 6285-57-0 | C7H5N3O2S | 195.2 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 6285-57-0
CAS Min % 96.0
CAS Max % 100.0
Melting Point 247°C to 252°C
Assay Percent Range 95% min. (HPLC)
Packaging Plastic bottle
Molecular Formula C7H5N3O2S
MDL Number MFCD00005786
Quantity 25 g
Beilstein 27, II, 232
Synonym 2-amino-6-nitrobenzothiazole, 6-nitrobenzo d thiazol-2-amine, 2-benzothiazolamine, 6-nitro, 6-nitrobenzothiazol-2-amine, 6-nitro-2-aminobenzothiazole, 2-amino-6-nitro benzothiazole, 6-nitro-2-benzothiazolamine, benzothiazole, 2-amino-6-nitro, ccris 1391, 6-nitro-benzothiazol-2-ylamine
InChI Key GPNAVOJCQIEKQF-UHFFFAOYSA-N
SMILES C1=CC2=C(C=C1[N+](=O)[O-])SC(=N2)N
IUPAC Name 6-nitro-1,3-benzothiazol-2-amine
Molecular Weight (g/mol) 195.2
PubChem CID 22704
Formula Weight 195.2
Percent Purity 96%
Chemical Name or Material 2-Amino-6-nitrobenzothiazole, 97%
Show More Show Less
compliance-icons
Product Identifier
  • 2-Amino-6-nitrobenzothiazole
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.