missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Amino-8-[(4-bromophenyl)azo]-1,7-dihydro-6H-purin-6-one-13D2,15N, TRC
Click to view available options
Quantity:
0.5 mg
1 mg
2.5 mg
Unit Size:
0.5mg
1mg
2.5mg
Specifications
Specifications
| Chemical Name or Material | 2-Amino-8-[(4-bromophenyl)azo]-1,7-dihydro-6H-purin-6-one-13C2,15N |
| Molecular Formula | 13C2 C9 H8 Br 15N N6 O |
| Quantity | 0.5 mg |
| Synonym | NSC 336665-13C2,15N, 2-Amino-8-[2-(4-bromophenyl)diazenyl]-1,9-dihydro-6H-purin-6-one-13C2,15N |
| InChI Formula | InChI=1S/C11H8BrN7O/c12-5-1-3-6(4-2-5)18-19-11-14-7-8(16-11)15-10(13)17-9(7)20/h1-4H,(H4,13,14,15,16,17,20)/b19-18+/i7+1,8+1,16+1 |
| SMILES | NC1=N[13c]2[15nH]c(\N=N\c3ccc(Br)cc3)n[13c]2C(=O)N1 |
| IUPAC Name | 2-amino-8-[(E)-(4-bromophenyl)diazenyl]-1,9-dihydropurin-6-one |
| Molecular Weight (g/mol) | 337.11 |
| Formula Weight | 336.001 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction