missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-9-((1S,3R,4S)-4-hydroxy-3-(hydroxymethyl)-2-methylenecyclopentyl)-1,9-dihydro-6H-purin-6-one hydrate, 98%, BLD Pharm™

Product Code. 31074304
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 mg
100 mg
250 mg
1 g
5 g
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
31074304 100 mg
31060557 250 mg
31062723 50 mg
31062266 5 g
31060566 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 31074304 Supplier BLD Pharm Supplier No. BD127328100mg
 Restricted Item
Estimated Shipment: 27-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 209216-23-9
Molecular Formula C12H17N5O4
MDL Number MFCD09754448
Quantity 100 mg
SMILES O=C1NC(N)=NC2=C1N=CN2[C@@H]3C([C@H](CO)[C@@H](O)C3)=C.[H]O[H]
Molecular Weight (g/mol) 295.29
Percent Purity 98%
Chemical Name or Material 2-Amino-9-((1S,3R,4S)-4-hydroxy-3-(hydroxymethyl)-2-methylenecyclopentyl)-1,9-dihydro-6H-purin-6-one hydrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.