missing translation for 'onlineSavingsMsg'
Learn More

2-Aminobenzenesulphonic Acid, TRC

Product Code. 30309463
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30309463 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30309463 Supplier Toronto Research Chemicals Supplier No. TRCA6225485G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Aniline-2-sulfonic acid
CAS 88-21-1
Molecular Formula C6 H7 N O3 S
Quantity 5 g
Synonym Benzenesulfonic acid, o-amino- (6CI,7CI,8CI), 2-Aminobenzenesulfonic acid, 1-Aminobenzene-2-sulfonic acid, 2-Aminophenylsulfonic acid, 2-Anilinesulfonic acid, 2-Sulfoaniline, Aniline-o-sulfonic acid, NSC 147, Orthanilic acid, o-Aminobenzenesulfonic acid, o-Aminophenylsulfonic acid, o-Sulfanilic acid
InChI Formula InChI=1S/C6H7NO3S/c7-5-3-1-2-4-6(5)11(8,9)10/h1-4H,7H2,(H,8,9,10)
SMILES Nc1ccccc1S(=O)(=O)O
IUPAC Name 2-aminobenzenesulfonic acid
Molecular Weight (g/mol) 173.19
Formula Weight 173.0147
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.