missing translation for 'onlineSavingsMsg'
Learn More

2-Aminobenzonitrile, TRC

Product Code. 30321407
Change view
Click to view available options
Quantity:
10 g
Unit Size:
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30321407 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30321407 Supplier Toronto Research Chemicals Supplier No. TRCA59170010G
 Restricted Item
Estimated Shipment: 27-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2-Aminobenzonitrile
CAS 1885-29-6
Molecular Formula C7H6N2
Quantity 10 g
InChI Formula InChI=1S/C7H6N2/c8-5-6-3-1-2-4-7(6)9/h1-4H,9H2
SMILES Nc1ccccc1C#N
IUPAC Name 2-aminobenzonitrile
Molecular Weight (g/mol) 118.14
Formula Weight 118.0531
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.