missing translation for 'onlineSavingsMsg'
Learn More

2-Aminobenzyl Alcohol, TRC

Product Code. 30315072
Change view
Click to view available options
Quantity:
50 g
Unit Size:
50g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30315072 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30315072 Supplier Toronto Research Chemicals Supplier No. TRCA60182850G
 Restricted Item
Estimated Shipment: 20-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material (2-Aminophenyl)methanol
CAS 5344-90-1
Molecular Formula C7 H9 N O
Quantity 50 g
Synonym (2-Aminophenyl)methanol, Mianserin Hydrochloride Imp. C (EP), Benzenemethanol, 2-amino-, Benzyl alcohol, o-amino- (6CI,7CI,8CI), 2-Aminobenzenemethanol, (o-Aminophenyl)methanol, 2-(Hydroxymethyl)aniline, 2-Aminobenzyl alcohol, NSC 1173, [2-(Hydroxymethyl)phenyl]amine, o-(Hydroxymethyl)aniline, o-Aminobenzyl alcohol, o-Aminobenzylic alcohol
InChI Formula InChI=1S/C7H9NO/c8-7-4-2-1-3-6(7)5-9/h1-4,9H,5,8H2
SMILES Nc1ccccc1CO
IUPAC Name (2-aminophenyl)methanol
Molecular Weight (g/mol) 123.15
Formula Weight 123.0684
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.