missing translation for 'onlineSavingsMsg'
Learn More

2-Aminoethyl 2-Aminoethanethiosulfonate Dihydrochloride, TRC

Product Code. 30322551
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30322551 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30322551 Supplier Toronto Research Chemicals Supplier No. TRCA608825100MG
 Restricted Item
Estimated Shipment: 12-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2-Aminoethyl 2-Aminoethanethiosulfonate, Dihydrochloride
CAS 10027-70-0
Molecular Formula C4 H12 N2 O2 S2 . 2 Cl H
Quantity 100 mg
InChI Formula InChI=1S/C4H12N2O2S2.2ClH/c5-1-3-9-10(7,8)4-2-6;;/h1-6H2;2*1H
SMILES Cl.Cl.NCCSS(=O)(=O)CCN
IUPAC Name 2-(2-aminoethylsulfonylsulfanyl)ethanamine;dihydrochloride
Molecular Weight (g/mol) 257.2
Formula Weight 255.9874
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.