missing translation for 'onlineSavingsMsg'
Learn More

2-Aminothiazole-4-carboxamide (~15% Inorganics), TRC

Product Code. 30136694
Change view
Click to view available options
Quantity:
50 mg
100 mg
500 mg
Unit Size:
100mg
500mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30136694 100 mg 100mg
30133051 500 mg 500mg
30004398 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30136694 Supplier Toronto Research Chemicals Supplier No. TRCA633288100MG
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 118452-02-1
Molecular Formula C4 H5 N3 O S
Molecular Weight (g/mol) 143.17
Synonym 2-Amino-4-thiazolecarboxamide, 2-Aminothiazole-4-carboxylic Acid Amide, NSC 85079
IUPAC Name 2-aminothiazole-4-carboxamide
SMILES NC(=O)c1csc(N)n1

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C4H5N3OS/c5-3(8)2-1-9-4(6)7-2/h1H,(H2,5,8)(H2,6,7)
Formula Weight 143.0153
Chemical Name or Material 2-Aminothiazole-4-carboxamide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.