missing translation for 'onlineSavingsMsg'
Learn More

2-Benzimidazolinone, Dr. Ehrenstorfer

Product Code. 30317330
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30317330 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30317330 Supplier Dr. Ehrenstorfer Supplier No. DREC10536550
 Restricted Item
Estimated Shipment: 16-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 1,3-Dihydro-2H-benzimidazol-2-one
CAS 615-16-7
Analyte or Component Names 1,3-Dihydro-2H-benzimidazol-2-one
Shipping Condition +20°C
Molecular Formula C7 H6 N2 O
Certifications/Compliance ISO/IEC 17025
Synonym Lansoprazole Imp. D (EP), Rabeprazole Sodium Imp. K (EP), Rabeprazole Imp. K (EP), 2H-Benzimidazol-2-one, 1,3-dihydro-, 2-Benzimidazolinone (6CI,7CI,8CI), 1,3-Dihydro-2H-benzimidazol-2-one, 1,3-Dihydro-2H-benzimidazole-2-one, 1,3-Dihydrobenzimidazol-2-one, 1H-Benzimidazol-2(3H)-one, 1H-Benzo[d]imidazol-2(3H)-one, 2(3H)-Benzimidazolone, 2(3H)-Oxobenzimidazole, 2,3-Dihydro-2-oxo-1H-benzimidazole, 2-Benzimidazolol, 2-Benzimidazolone, 2-Hydroxybenzimidazole, N,N'-(1,2-Phenyleneurea), NSC 10383, NSC 178108, Urea, N,N'-(1,2-phenylene)-, o-Phenyleneurea, Lansoprazole Impurity D, Rabeprazole Sodium Impurity K, Rabeprazole Impurity K
InChI Formula InChI=1S/C7H6N2O/c10-7-8-5-3-1-2-4-6(5)9-7/h1-4H,(H2,8,9,10)
SMILES O=C1Nc2ccccc2N1
IUPAC Name 1,3-dihydrobenzimidazol-2-one
Molecular Weight (g/mol) 134.14
Quantity 100 mg
Formula Weight 134.048 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.