missing translation for 'onlineSavingsMsg'
Learn More

2-Benzothiazolethiol, TRC

Product Code. 30343980
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30343980 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30343980 Supplier Toronto Research Chemicals Supplier No. TRCB2066505G

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2-Mercaptobenzothiazole
CAS 149-30-4
Molecular Formula C7H5NS2
Quantity 5 g
Synonym 2(3H)-Benzothiazolethione, 2-Benzothiazolethiol (8CI), 2-Benzothiazolinethione (6CI,7CI), 1,3-Benzothiazole-2-thiol, 1,3-Benzothiazole-2-thione, 2,3-Dihydrobenzothiazole-2-thione, 2-Benzothiazolyl mercaptan, 2-MBT, 2-Mercapto-1H-benzothiazole, 2-Mercaptobenzothiazole, 2-Mercaptobenzthiazole, 2-Sulfanylbenzothiazole, Accel M, Accelerator M, Aero 407, Aero Promoter 412, Benz-1,3-thiazolidine-2-thione, Benzo[d]thiazole-2-thiol, Benzothiazolethiol, Captax, Captax MBT, DT 402, Dermacid, Ekagom G, Kaptaks, Kaptax, M 2, M 2 (rust inhibitor), MBT, MBT (vulcanization accelerator), Mebetizol, Mebetizole, Mebithizol, Mercaptobenzothiazole, Mercaptobenzthiazole, Mertax, NSC 2041, Nocceler M, Nocceler M-P, Nonflex NB, Nuodeb 84, OCID, Oricel M, Perkacit MBT, Pneumax MBT, RU 3, Rotax, Royal MBT, Sanceler M, Sanceler M-G, Soxinol M, Thiotax, Vulkacit M, Vulkacit Mercapto, Vulkacit Mercapto MG/C, Vulkacit Mercapto/C, Vulkacit Mercapto/MG, Vulkafil ZN 94TT01, Wobezit M, accelerator MBT
InChI Formula InChI=1S/C7H5NS2/c9-7-8-5-3-1-2-4-6(5)10-7/h1-4H,(H,8,9)
SMILES Sc1nc2ccccc2s1
IUPAC Name 1,3-benzothiazole-2-thiol
Molecular Weight (g/mol) 167.2513
Formula Weight 166.9863
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.