missing translation for 'onlineSavingsMsg'
Learn More

2-Benzoylnaphthalene, 98%

Product Code. 11419286
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11419286 1 g 1g
11429286 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11419286 Supplier Thermo Scientific Alfa Aesar Supplier No. A18121.03
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 644-13-3 | C17H12O | 232.28 g/mol

2-Benzoylnaphthalene is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Benzoylnaphthalene is used as pharmaceutical intermediate.

Solubility
Soluble in hot methanol.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 644-13-3
Molecular Formula C17H12O
Molecular Weight (g/mol) 232.28
MDL Number MFCD00004106
InChI Key SJNXJRVDSTZUFB-UHFFFAOYSA-N
Synonym 2-benzoylnaphthalene, 2-naphthyl phenyl ketone, 2-benzonaphthone, naphthalen-2-yl phenyl methanone, methanone, 2-naphthalenylphenyl, 2'-benzonaphthone, ketone, 2-naphthyl phenyl, 2-naphthylphenylketone, beta-benzoylnaphthalene, 2-naphthyl phenyl methanone
PubChem CID 69516
IUPAC Name naphthalen-2-yl(phenyl)methanone
SMILES O=C(C1=CC=CC=C1)C1=CC=C2C=CC=CC2=C1

Specifications

Melting Point 80°C to 82°C
Boiling Point 216°C to 216°C (7 mmHg)
Quantity 1 g
Solubility Information Soluble in hot methanol.
Formula Weight 232.28
Percent Purity 98%
Chemical Name or Material 2-Benzoylnaphthalene

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.