missing translation for 'onlineSavingsMsg'
Learn More

2-(Benzyloxy)phenol, TRC

Product Code. 30338479
Change view
Click to view available options
Quantity:
5 g
10 g
Unit Size:
10g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30338479 5 g 5g
30347392 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30338479 Supplier Toronto Research Chemicals Supplier No. TRCB2866555G
 Restricted Item
Estimated Shipment: 06-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2-(Benzyloxy)phenol
CAS 6272-38-4
Molecular Formula C13H12O2
Quantity 5 g
Synonym Benzyl o-Hydroxyphenyl Ether, NSC 37426, NSC 72669, o-(Benzyloxy)phenol
InChI Formula InChI=1S/C13H12O2/c14-12-8-4-5-9-13(12)15-10-11-6-2-1-3-7-11/h1-9,14H,10H2
SMILES Oc1ccccc1OCc2ccccc2
IUPAC Name 2-phenylmethoxyphenol
Molecular Weight (g/mol) 200.23
Formula Weight 200.0837
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.