missing translation for 'onlineSavingsMsg'
Learn More

2-Benzyloxypyridine-3-boronic acid, 95%, Thermo Scientific™

Product Code. 15473577
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
This item is not returnable. View return policy

Product Code. 15473577

Brand: Thermo Scientific Alfa Aesar H26978.MD

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1072952-41-0
Molecular Formula C12H12BNO3
Molecular Weight (g/mol) 229.042
MDL Number MFCD06798264
InChI Key YTIGWZQYGHOXOB-UHFFFAOYSA-N
Synonym 2-benzyloxypyridine-3-boronic acid, 2-benzyloxy pyridin-3-yl boronic acid, 2-benzyloxy pyridin-3-ylboronic acid, 2-benzyloxy pyridine-3-boronic acid, acmc-2098up, 2-benzyloxy-3-pyridine boronic acid, 2-benzyloxy-pyridin-3-yl boronic acid, 2-benzyloxy pyridin-3-yl boronicacid, 2-benzyloxypyridin-3-ylboronic acid
PubChem CID 44755161
IUPAC Name (2-phenylmethoxypyridin-3-yl)boronic acid
SMILES B(C1=C(N=CC=C1)OCC2=CC=CC=C2)(O)O

Specifications

Melting Point 145°C to 147°C
Quantity 250 mg
Formula Weight 229.04
Percent Purity 95%
Chemical Name or Material 2-Benzyloxypyridine-3-boronic acid
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.