Learn More
2-Biphenylisocyanate, TRC
Specifications
Specifications
| Chemical Name or Material | 2-Biphenylisocyanate |
| CAS | 17337-13-2 |
| Molecular Formula | C13H9NO |
| Quantity | 1 g |
| InChI Formula | InChI=1S/C13H9NO/c15-10-14-13-9-5-4-8-12(13)11-6-2-1-3-7-11/h1-9H |
| SMILES | O=C=Nc1ccccc1c2ccccc2 |
| IUPAC Name | 1-isocyanato-2-phenylbenzene |
| Molecular Weight (g/mol) | 195.21 |
| Formula Weight | 195.0684 |