missing translation for 'onlineSavingsMsg'
Learn More

2-Biphenylmethanol, 98%

Product Code. 11347136
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11347136 5 g 5g
11337136 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11347136 Supplier Thermo Scientific Alfa Aesar Supplier No. L07094.06
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 2928-43-0 | C13H12O | 184.238 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2928-43-0
Molecular Formula C13H12O
Molecular Weight (g/mol) 184.238
MDL Number MFCD00004620
InChI Key VKTQADPEPIVMHK-UHFFFAOYSA-N
Synonym 2-biphenylmethanol, 1,1'-biphenyl-2-ylmethanol, 2-phenylphenyl methanol, biphenylmethanol, 2-phenylbenzyl alcohol, 1,1'-biphenyl methanol, biphenyl-2-yl-methanol, 2-phenylphenyl methan-1-ol, 1,1'-biphenyl-2-methanol, biphenyl-2-ylmethanol
PubChem CID 76229
IUPAC Name (2-phenylphenyl)methanol
SMILES C1=CC=C(C=C1)C2=CC=CC=C2CO

Specifications

Melting Point 47°C to 51°C
Boiling Point 96°C (0.04 mmHg)
Quantity 5 g
Beilstein 1866883
Formula Weight 184.24
Percent Purity 98%
Chemical Name or Material 2-Biphenylmethanol

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.